Learn More
3M™ Lexa™ Safety Glasses 15236
Click to view available options
Lens Tint: Gray
Description
- Aspheric lens provides expansive coverage and a virtually unobstructed viewing range
- Frame is fully customizable with different lens angles and temple lengths
- DX coating provides enhanced anti-scratch and anti-fog protection
- Polycarbonate lens absorbs 99.9%UV
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses |
Frame Color | Black |