Learn More
Levulinic Acid, 98+%, Thermo Scientific Chemicals
Catalog Number p-3736790
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
123-76-2
C5H8O3
116.12
MFCD00002796
JOOXCMJARBKPKM-UHFFFAOYSA-N
levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid
11579
CHEBI:45630
4-oxopentanoic acid
CC(=O)CCC(=O)O
123-76-2
C5H8O3
116.12
MFCD00002796
JOOXCMJARBKPKM-UHFFFAOYSA-N
levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid
11579
CHEBI:45630
4-oxopentanoic acid
CC(=O)CCC(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 33.0°C to 35.0°C
Color Yellow
Density 1.1300g/mL
Boiling Point 245.0°C to 246.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3COCH2CH2CO2H
Refractive Index 1.4396 to 1.4426
Quantity 100 g
Beilstein 03, 671
Fieser 01,564; 02,239; 10,230
Merck Index 15, 5526
Specific Gravity 1.13
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and ether,insoluble in aliphatic hydrocarbons
Formula Weight 116.12
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material Levulinic acid
Show More Show Less
compliance-icons
Product Identifier
  • Levulinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.