Learn More
BRAND™ LDPE Narrow Neck Bottle for Schilling Burettes
48.68 NOK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Use promo code "23264" to get your promotional price.
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Description
- Low-density Polyethylene (LDPE) construction
- Narrow mouth resevoir bottle
- With tube bushing
- For use with Schilling burettes
Specifications
Product Type | Reservoir Bottle |
Capacity (Metric) | 500 mL |
Material | LDPE |