missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (555) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5674JF585
This item is not returnable.
View return policy
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| L1CAM | |
| Monoclonal | |
| Janelia Fluor 585 | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Mouse cerebellum-derived partially purified L1CAM | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 555 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 3897 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction