missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Glutamic acid 5-cyclohexyl ester, 95%

Brand:  Alfa Aesar™ H63616.06

Additional Details : CAS Number : 112471-82-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430409

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

112471-82-6
229.276
JSYMWKYNCWQUOY-VIFPVBQESA-N
14283856
C1CCC(CC1)OC(=O)CCC(C(=O)O)N
C11H19NO4
MFCD00153436
h-glu ochex-oh, h-glu ochx-oh, l-glutamic acid 5-cyclohexyl ester, s-2-amino-5-cyclohexyloxy-5-oxopentanoic acid, pubchem14922, l-glutamic acid gamma-cyclohexyl ester, 2s-2-amino-5-cyclohexyloxy-5-oxopentanoic acid
(2S)-2-amino-5-cyclohexyloxy-5-oxopentanoic acid

Specifications

L-Glutamic acid 5-cyclohexyl ester
C11H19NO4
h-glu ochex-oh, h-glu ochx-oh, l-glutamic acid 5-cyclohexyl ester, s-2-amino-5-cyclohexyloxy-5-oxopentanoic acid, pubchem14922, l-glutamic acid gamma-cyclohexyl ester, 2s-2-amino-5-cyclohexyloxy-5-oxopentanoic acid
C1CCC(CC1)OC(=O)CCC(C(=O)O)N
229.276
229.27
95%
112471-82-6
MFCD00153436
JSYMWKYNCWQUOY-VIFPVBQESA-N
(2S)-2-amino-5-cyclohexyloxy-5-oxopentanoic acid
14283856
Solid
5g
Videos
SDS Documents