Learn More
L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™
Catalog Number p-7058160
Click to view available options
Quantity:
250 mg
1 g
5 g
39825-33-7
C6H13NO2
131.18
MFCD08059709,MFCD08059709,MFCD12796134
QDQVXVRZVCTVHE-UHFFFAOYNA-N
s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
13089460
propan-2-yl 2-aminopropanoate
CC(C)OC(=O)C(C)N
39825-33-7
C6H13NO2
131.18
MFCD08059709,MFCD08059709,MFCD12796134
QDQVXVRZVCTVHE-UHFFFAOYNA-N
s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
13089460
propan-2-yl 2-aminopropanoate
CC(C)OC(=O)C(C)N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Formula Weight 167.63
Percent Purity 95%
Quantity 250 mg
Chemical Name or Material L-Alanine isopropyl ester hydrochloride