Learn More
L-Alanine-2-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | L-Alanine-2-D |
| CAS | 21386-65-2 |
| Molecular Formula | C3 D H6 N O2 |
| Quantity | 0.5 g |
| Synonym | L-Alanine-2-d (9 CI, ACI), Alanine-2-d (6 CI, 8 CI), (2 S)-2-Amino-2-deuteriopropanoic acid, L-(α2H)Alanine, (2 S)-2-Aminopropanoic-2-d acid, L-Alanine-2-D, L-[2-2H1]Alanine |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i2D |
| SMILES | [2 H][C@@](C)(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2-deuteriopropanoic acid |
| Molecular Weight (g/mol) | 90.0993 |
| Formula Weight | 90.054 g/mol |
| Show More |