Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint
Ideal for use with rotary evaporators
676.03 NOK - 2296.23 NOK
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Equipment) | Rotary evaporator |
Neck Style | Single |
Product Code | Brand | Capacity (Metric) | For Use With (Application) | Standard Taper Joint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | For Use With (Application) | Standard Taper Joint | Price | Quantity & Availability | |||
11802933
|
DWK Life Sciences
6086754224 |
100 mL | Square bead joints provide superior clamping to standard Joint beads | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11892923
|
DWK Life Sciences
6086754624 |
500 mL | Square bead joints provide superior clamping to standard Joint beads | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11707949
|
DWK Life Sciences
KC2516550 |
50 mL | Plastic coated Glassware provides an added measure of safety in the event of breakage | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11717949
|
DWK Life Sciences
KC25165100 |
100 mL | Plastic coated Glassware provides an added measure of safety in the event of breakage | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11727949
|
DWK Life Sciences
KC25165200 |
200 mL | Plastic coated Glassware provides an added measure of safety in the event of breakage | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11747949
|
DWK Life Sciences
KC251651000 |
1000 mL | Plastic coated Glassware provides an added measure of safety in the event of breakage | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11737949
|
DWK Life Sciences
KC25165500 |
500 mL | Plastic coated Glassware provides an added measure of safety in the event of breakage | 24/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Ideal for use with rotary evaporators
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- Single-neck flask with standard taper outer joint
- Designed for easy recovery of reaction products
- Total contents are accessible with a spatula or policeman
- Lower portion fits standard heating mantles
- Even wall thickness and minimum runout
Specifications
Borosilicate Glass | |
Rotary evaporator | |
Single |