Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Ferrule for 1/16 in. Tubing
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
Brand: DWK Life Sciences 420822-0116
51.35 NOK valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- EFTE Flangeless Ferrule for 1/16 in. OD Tubing
Specifications
For 0.06 in. O.D. Tubing and Chromaflex Column Chromatography Fitting | |
ETFE | |
Ferrule | |
Case of 1 |