Learn More
Kimberly-Clark™ Professional Kimtech Science™ Comfort Nitrile Gloves
Experience the feeling of high comfort, combined with quality. Kimtech Science™ Comfort Nitrile Gloves are specifically engineered for protection in low risk research applications.
Brand:  Kimberly-Clark™ Professional 47676
84.54 NOK valid until 2018-08-31
Use promo code "13204" to get your promotional price.
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15362206
  • 94.99 NOK / Pack of 150
In Stock 14
Add to basket
Description
Description
  • Combines high comfort, quality and cleanliness for protection in low risk research applications
  • A brand-new nitrile formulation engineered for comfort
  • 150 gloves per standard sized box for all sizes
  • Better chemical protection than latex, tested on the 6 most common chemicals in the lab
  • EPI Category III
Specifications
Specifications
Nitrile
150/Box
White
X-Large
Powder-free
Videos
Documentation Special Offers
Special Offers
  • Kimberly Clark
    Comfort and fitting are main criteria for End user to adopt a PPE and use it properly
  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13204
    2018-08-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 04/06/2018 and 31/08/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions