missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55407-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly.Specifications
KIF15 | |
Polyclonal | |
Unconjugated | |
KIF15 | |
HKLP2, KIF15, kinesin family member 15, Kinesin like protein 2, Kinesin like protein 7, Kinesin like protein KIF15, KLP2, KNSL7, NY BR 62 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209737, 353302, 56992 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9L6, Q7TSP2, Q9NS87 | |
Kif15 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |