missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM4D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22591-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.Specifications
KDM4D | |
Polyclonal | |
Unconjugated | |
KDM4D | |
JHDM3D, JMJD2D, KDM4D, lysine (K) demethylase 4D, Lysine specific demethylase 4D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55693 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6B0I6 | |
KDM4D | |
KDM4D Fusion Protein Ag18436 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |