Learn More
Abnova™ JAK2/CEN9q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0184.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16003295
  • 29855.00 NOK / 200µL
Estimated Shipment: 29-05-2024
to see stock.
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Store at 4°C in the dark.
Tube
3717
200 μL
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe