Learn More
Invitrogen™ Human JAK1, GST Tag Recombinant Protein
Janus kinase 1 (JAK1) is a protein tyrosine kinase.
5140.00 NOK - 112500.00 NOK
Specifications
Accession Number | NP_002218.2 |
---|---|
Gene Alias | JAK1, JAK1A, JAK1B |
KinaseFamily | JAK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438753
|
Invitrogen™
PV4775 |
1mg |
112500.00 NOK
1mg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10748585
|
Invitrogen™
PV4774 |
10 μg |
5140.00 NOK
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488683
|
Invitrogen™
PR8767C |
100μg |
17740.00 NOK
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
JAK1 is involved in the interferon-alpha/beta and -gamma signal transduction pathways.
Specifications
NP_002218.2 | |
JAK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
JAK1, JAK1A, JAK1B | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Cytoplasmic, Catalytic | |
Signal Transduction |