missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropylboronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Quantity | 1 g |
Beilstein | 1731883 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 87.91 |
Percent Purity | 98% |
Chemical Name or Material | Isopropylboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'