Learn More
isopropyl 2-bromobenzoate, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
100 g
Specifications
Assay Percent Range | 97+% |
Quantity | 1 g |
Synonym | isopropyl 2-bromobenzoate, isopropyl o-bromobenzoate, methylethyl 2-bromobenzoate, prop-2-yl 2-bromobenzoate, isopropyl 2-bromobenzoate #, isopropyl2-bromobenzoate, 2-bromobenzoic acid isopropyl ester, 2-bromobenzoic acid, isopropyl ester, benzoic acid, 2-bromo-,1-methylethyl ester, benzoic acid, 2-bromo-, 1-methylethyl ester |
Formula Weight | 243.1 |
Percent Purity | 97% |
Chemical Name or Material | Isopropyl 2-bromobenzoate |