Learn More
Isobutyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog Number p-7024379
Click to view available options
Quantity:
5 g
25 g
100 g
591-82-2
C5H9NS
115.194
MFCD00022062
NSDDRJXKROCWRZ-UHFFFAOYSA-N
isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
68960
1-isothiocyanato-2-methylpropane
CC(C)CN=C=S
591-82-2
C5H9NS
115.194
MFCD00022062
NSDDRJXKROCWRZ-UHFFFAOYSA-N
isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
68960
1-isothiocyanato-2-methylpropane
CC(C)CN=C=S
It is isothiocyanate compound used for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is isothiocyanate compound used for proteomics research.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Density 0.94
Boiling Point 160°C to 162°C
Flash Point 24°C (75°F)
Odor Pungent
Refractive Index 1.498
Quantity 5 g
UN Number UN2924
Beilstein 1740371
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material Isobutyl isothiocyanate