missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ISL2 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898554
|
Proteintech
18659-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16888554
|
Proteintech
18659-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.Specifications
ISL2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P50480, Q96A47, Q9CXV0 | |
104360, 57233, 64843 | |
ISL2 Fusion Protein Ag13333 | |
Primary | |
-20°C | |
ISL2 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ISL LIM homeobox 2, ISL2, Islet 2 | |
Isl2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |