missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
424.82 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >100°C (decomposition) |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1481 |
Solubility Information | Soluble in alcohol and water (?98g/100 mL H2O at 0?°C). |
Formula Weight | 254.75 (Anhydrous) |
Grade | Reagent |
Sensitivity | Hygroscopic |
Chemical Name or Material | Iron(II) perchlorate hydrate |
Safety and Handling
Product Identifier
- Iron(II) perchlorate hydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'