missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) iodide, anhydrous, 97% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048789
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7783-86-0
MFCD00016094
iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis
This item is not returnable. View return policy
7783-86-0
MFCD00016094
iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis

Iron(II) iodide is used as a catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Physical Form Crystalline
Quantity 2 g
UN Number UN3260
Solubility Information Soluble in water,diethyl ether and ethanol.
Formula Weight 309.66
Percent Purity 97%
Odor Odorless
Sensitivity Air and light sensitive; Hygroscopic
Density 5.32 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) iodide, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) iodide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'