missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide hydrate, Thermo Scientific Chemicals

Catalog Number p-7049914
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
13463-12-2
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
λ²-iron(2+) dibromide
[Fe++].[Br-].[Br-]
This item is not returnable. View return policy
13463-12-2
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
λ²-iron(2+) dibromide
[Fe++].[Br-].[Br-]

It is widely as a pharmaceutical intermediate.It is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 27°C
Color Green
Physical Form Lump
Quantity 25 g
Linear Formula FeBr2·xH2O
UN Number UN3260
Solubility Information It is soluble in water,soluble in other solvents such as THF,methanol,ethanol.
Formula Weight 215.67 (Anhydrous)
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.9 g/mL
Chemical Name or Material Iron(II) bromide hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) bromide hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'