missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (PCRP-IRF3-3B2) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14106JF669
5261.50 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| IRF3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| interferon regulatory factor 3, IRF-3 | |
| Recombinant full-length human IRF3 protein (Uniprot: Q14653) | |
| 0.1 mL | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-IRF3-3B2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction