Learn More
BINDER™ IQ/OQ Qualification Documents
IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists including calibration guide and comprehensive unit documentation. For APT-COM GLP software.
17980.00 NOK - 20440.00 NOK
Specifications
For Use With (Equipment) | Multi Management Software APT-COM™ GLP-Edition Software |
---|---|
Product Line | APT-COM™ |
dimServiceOffering | GxP Compliant IQ/OQ Documentation |
For Use With (Application) | Installation and Operational Qualification |
Includes | IQ/OQ Checklists with Calibration Guide and Comprehensive Unit Documentation |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
16414019
|
BINDER™
7057-0004 |
IQ/OQ Documentation (PDF) |
17980.00 NOK
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16444009
|
BINDER™
7007-0004 |
IQ/OQ Documentation (Hard Copy) |
20440.00 NOK
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Installation and operational qualification documents for validation performed by customers
- Designed for use with APT-COM™ GLP-Edition software
- Includes IQ/OQ checklists, calibration guide, and comprehensive unit documentation
- Both digital and hard copy options are available
Specifications
Multi Management Software APT-COM™ GLP-Edition Software | |
GxP Compliant IQ/OQ Documentation | |
IQ/OQ Checklists with Calibration Guide and Comprehensive Unit Documentation |
APT-COM™ | |
Installation and Operational Qualification |