Learn More
Invitrogen™ S.N.A.P.™ Plasmid DNA MiniPrep Kit
The S.N.A.P.™ MiniPrep Kit is plasmid DNA purification system designed to yield pure plasmid DNA for automated sequencing in just 25 minutes.
Brand: Invitrogen™ K190001
Additional Details : Weight : 0.56000kg
Includes: S.N.A.P. MidiPrep filtering columns, S.N.A.P. MidiPrep DNA binding columns, lysis buffer, binding buffer, wash buffer, 4X final wash, resuspension buffer, precipitation salt, and RNase A
Description
The S.N.A.P.™ MiniPrep Kit is plasmid DNA purification system designed to yield pure plasmid DNA (Figures 1 and 2) for automated sequencing in just 25 minutes
- DNA can improve your automated sequencing results by producing:
- Longer reads
- Lower background
- Higher accuracy
- Stronger signal peaks
Specifications
Store the S.N.A.P.™ MiniPrep spin columns, lysis buffer, binding buffer, wash buffer, 4X final wash, and the collection tubes at room temperature. Store the resuspension buffer, precipitation salt solution, and RNase A at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
Spin Column | |
Plasmid DNA | |
Not High-throughput Compatible (Manual) | |
< 100 μg (Small-Scale) Plasmid DNA | |
Plasmid DNA MiniPrep Kit | |
Plasmid DNA |
Kit | |
Bacterial Culture | |
Next-Generation Sequencing, In Vitro Transcription, Sequencing, Cloning, Transformation | |
100 Preps | |
S.N.A.P. ™ | |
100 Preps | |
25 min. |
For Research Use Only. Not for use in diagnostic procedures.