Learn More
Invitrogen™ PureLink™ HiPure Plasmid Filter Midi/Maxi Starter Kit
Purify high-quality, transfection-grade plasmid DNA in less than two hours
Brand: Invitrogen™ 114756S
Additional Details : Weight : 0.10000kg
Includes: 4 columns
Description
Pure Link HiPure Filter Plasmid Kits are designed to isolate plasmid DNA of highest purity from 25 to 500mLs of bacterial cultures. The PureLink HiPure Plasmid Filter Kits include HiPure Filter Columns, which combine filtration for clarification of bacterial lysate with anion-exchange resin purification of plasmid DNA, in a single unit. The resulting plasmid DNA is of quality equivalent to that obtained by purification with two passes through cesium chloride gradients - the most rigorous method for DNA purification. For transfections and other applications with no need for additional steps to remove contaminants like RNA, proteins, or endotoxins. Additionally, the elimination of phenol, chloroform, ethidium bromide, and cesium chloride from the protocol minimizes exposure to, and disposal of hazardous materials.
- PureLink HiPure Plasmid Filter Kits provide: Simple protocols - bacterial lysate clarification without centrifugation
- High yields - up to 200μg for midipreps and up to 850μg for maxipreps
- Reliable performance - High-quality plasmid DNA purified with these kits is suitable for wide variety of applications
- Addition of a HiPure Filter Column to the PureLink HiPure Plasmid Kits provides rapid clearing of the bacterial lysate without need for centrifugation
- Lysate filtration cartridge is integrated into the DNA-binding column for combined step of lysate clarification and plasmid DNA binding directly to anion-exchange resin
- A single column wash removes impurities such as RNA, proteins, metabolites, and other low-molecular-weight molecules, and the ultrapure plasmid DNA is eluted with a high-salt buffer
- DNA is then desalted and concentrated with isopropanol precipitation step, then collected by centrifugation
- Entire protocol can be completed in about two hours; purification time is 90 minutes
DNA and RNA Purification and Analysis, DNA Extraction, Midiprep, Plasmid DNA Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Anion Exchange Resin | |
Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation | |
100-200μg (Medium-Scale) Plasmid DNA | |
1000 μg |
Bacterial Culture | |
2 | |
Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.