missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 5 Antibody (P5H9), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2528Z
5138.25 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Integrin alpha V beta 5 Monoclonal antibody specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF
Specifications
| Integrin alpha V beta 5 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3685 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF | |
| P5H9 | |
| Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF-ready | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| HT1080 human fibrosarcoma cell line | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction