Learn More
Miele Professional™ Insert
Insert E 402 for optimum loading of up to 44 watch glasses
Brand: Miele Professional™ 3830420
Additional Details : Weight : 1.03400kg
Description
- Suitable for 44 watch glasses Ø 80 - 125 mm
- equipped with 23 bars spaced 15 mm apart
Specifications
Stainless Steel | |
Stainless Steel | |
Washer-disinfectors, medical | |
53 mm |
Insert/Module for Watch Glasses | |
44 watch glasses | |
445 mm | |
200 mm |