missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B Hemagglutinin 2 Antibody (InB190) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-73164JF635
This item is not returnable.
View return policy
Description
Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Influenza B Hemagglutinin 2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| InB190 | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza B virus. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction