missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-carboxylic acid, 98%, Thermo Scientific™

Catalog Number p-3778871
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2124-55-2
C9H7NO2
161.16
MFCD00009739
ROGHUJUFCRFUSO-UHFFFAOYSA-N
indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
595229
1H-indole-4-carboxylic acid
C1=CC(=C2C=CNC2=C1)C(=O)O
This item is not returnable. View return policy
2124-55-2
C9H7NO2
161.16
MFCD00009739
ROGHUJUFCRFUSO-UHFFFAOYSA-N
indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
595229
1H-indole-4-carboxylic acid
C1=CC(=C2C=CNC2=C1)C(=O)O
Melting Point 207°C to 209°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indole-4-carboxylic acid
compliance-icons
Product Identifier
  • Indole-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'