missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-propionic acid, ∽99%, MP Biomedicals™

Catalog Number p-4404412
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
This item is not returnable. View return policy
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
OC(=O)CCC1=CNC2=CC=CC=C12
This item is not returnable. View return policy
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
OC(=O)CCC1=CNC2=CC=CC=C12
Color Cream
Assay Percent Range ∽ 99%
Quantity 1 g
Formula Weight 189.21
Percent Purity ∽99%
Physical Form Powder
Chemical Name or Material Indole-3-propionic acid, Molecular biology
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'