missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric acid, MP Biomedicals™

Catalog Number p-4404411
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
133-32-4
C12H13NO2
203.241
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
This item is not returnable. View return policy
133-32-4
C12H13NO2
203.241
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
Color White
Quantity 1 g
Formula Weight 203.24
Physical Form Powder
Chemical Name or Material Indole-3-butyric acid, Plant growth hormone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'