missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) ethoxide, 99.9% (metals basis), Thermo Scientific™

Catalog Number p-7059482
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
62763-48-8
C6H15InO3
250.00
MFCD00799104
MCXZOLDSEPCWRB-UHFFFAOYSA-N
indium ethoxide, indium triethoxide
16701139
triethoxyindigane
CCO[In](OCC)OCC
This item is not returnable. View return policy
62763-48-8
C6H15InO3
250.00
MFCD00799104
MCXZOLDSEPCWRB-UHFFFAOYSA-N
indium ethoxide, indium triethoxide
16701139
triethoxyindigane
CCO[In](OCC)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 150°C (decomposition)
Color White
Linear Formula In(OCH2CH3)3
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 250
Percent Purity 99.9%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Indium (III) ethoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'