Learn More
Velp Scientifica™ Imhoff Cone
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
730.00 NOK - 2215.00 NOK
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
15379624
|
Velp Scientifica™
A00001003 |
Glass |
2215.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15369624
|
Velp Scientifica™
A00001002 |
Plastic |
730.00 NOK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
- Transparent for visability
- Available in glass or plastic
- Available in graduated
- For use with Flocculators
Specifications
1000 mL | |
FCS, JLT |