Learn More
IL-13 Antibody (tralokinumab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28679JF525
3217.25 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 30171651
  • 3790.00 NOK / 0.10mL
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-13
Monoclonal
Janelia Fluor 525
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
IL-13
0.1 mL
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
3596
Store at 4°C in the dark.
IgG4
Flow Cytometry, ELISA, Functional Assay
tralokinumab
50mM Sodium Borate
Human
Protein A purified