missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2 Human, DyLight 650, Isotype Control, Novus Biologicals™
Human Monoclonal Isotype Control
Brand: Bio-Techne NBP3-06870C
This item is not returnable.
View return policy
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for ControlSpecifications
IgG2 | |
MAB1641 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
3501 | |
Store at 4°C in the dark. | |
Purified |
Monoclonal | |
DyLight 650 | |
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
Human myeloma serum | |
0.1 mL | |
Immunology | |
Human | |
Isotype Controls | |
IgG2 |