missing translation for 'onlineSavingsMsg'
Learn More

IgG (H+L) F(ab′)2-Goat anti-Mouse, PerCP-eFluor™ 710, eBioscience™

F(ab′)2-Goat Polyclonal Secondary Antibody

Brand:  Affymetrix eBioscience 46-4010-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15589246

  • 806.00 NOK / 25µg


Estimated Shipment
08-09-2020
Add to basket

Description

Description

Description: The goat anti-mouse IgG polyclonal antibody reacts with the heavy and light chains of mouse IgG, with minimal cross reactivity to human, rat, bovine, goat, hamster, and rabbit serum proteins. Reactivity has been shown to IgG1, IgG2a, IgG2b, and IgG3, as well as some reactivity to IgM. The antibody is a F(ab')2 fragment which contains the 2 Fab domains still linked through the hinge region of the immunoglobulin. Removal of the Fc region helps to reduce issues typically found with Fc-mediated binding.This polyclonal has been validated as a secondary reagent (its ability to recognize cells stained with a mouse IgG followed by this polyclonal antibody).Applications Reported: This polyclonal antibody has been reported for use in flow cytometric analysis.Applications Tested: This polyclonal antibody has been tested by flow cytometric analysis of cells stained with a mouse IgG antibody. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG (H+L)
Polyclonal
PerCP-eFluor 710
Liquid
Ig
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Secondary
Murine
Flow Cytometry
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
F(ab')2-Goat
Affinity chromatography
RUO
Secondary Antibody
Polyclonal
Videos
Documents

For Research Use Only.