Learn More
Sino Biological™ IFNB1 Recombinant Mouse Protein, hIgG1-Fc Tag
Expressed from DNA sequence encoding extracellular domain of mouse IFNB1 (Met 1-Asn 182) fused to Fc region of human IgG1 at C-terminus
Brand: Sino Biological™ 50708M02H5
Additional Details : Weight : 0.01000kg
Description
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications
Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications | |
15977 | |
≥90% by SDS-PAGE | |
Mouse | |
<1 EU/μg | |
IFNB1 | |
Other Proteins | |
IFNB1 Recombinant Mouse Protein |
Lyophilized | |
46.7kd | |
5μg | |
Truncated, Recombinant | |
Interferon Beta 1, Fibroblast | |
Assay Standard | |
Mammalian | |
IgG1 Fc Tag |
For Research Use Only. Not for use in diagnostic procedures.