Learn More
Ibufenac Methyl Ester, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Ibufenac Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Ibufenac Methyl Ester |
| CAS | 61566-33-4 |
| Analyte or Component Names | Ibufenac Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(2-Methylpropyl)benzeneacetic acid methyl ester, 2-(4-Isobutylphenyl)acetic acid methyl ester, Methyl (4-isobutylphenyl)acetate |
| InChI Formula | InChI=1S/C13H18O2/c1-10(2)8-11-4-6-12(7-5-11)9-13(14)15-3/h4-7,10H,8-9H2,1-3H3 |
| SMILES | COC(=O)Cc1ccc(CC(C)C)cc1 |
| IUPAC Name | methyl 2-[4-(2-methylpropyl)phenyl]acetate |
| Show More |