missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iASPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18590-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody could both recognize unphosphorylated and phosphorylated iASPP.
This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter′s anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.Specifications
iASPP | |
Polyclonal | |
Unconjugated | |
PPP1R13L | |
IASPP, Inhibitor of ASPP protein, NKIP1, PPP1R13B like protein, PPP1R13BL, PPP1R13L, Protein iASPP, RAI, RelA associated inhibitor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10848, 333654, 686781 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5I1X5, Q8WUF5 | |
Ppp1r13l | |
iASPP Fusion Protein Ag13273 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |