Learn More
Hydroxyprogesterone Caproate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Hydroxyprogesterone Caproate
Specifications
Specifications
| Chemical Name or Material | Hydroxyprogesterone Caproate |
| CAS | 630-56-8 |
| Analyte or Component Names | Hydroxyprogesterone Caproate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H40 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Hydroxyprogesterone Caproate, Pregn-4-ene-3,20-dione, 17-hydroxy-, hexanoate (7CI,8CI), Progesterone, 17-hydroxy-, hexanoate (6CI), Hexanoic acid, ester with 17-hydroxypregn-4-ene-3,20-dione (8CI), 17α-Caproyloxy-P4, 17α-Caproyloxypregn-4-ene-3,20-dione, 17α-Caproyloxyprogesterone, 17α-Hydroxypregn-4-ene-3,20-dione caproate, 17α-Hydroxypregn-4-ene-3,20-dione hexanoate, 17α-Hydroxyprogesterone caproate, 17α-Hydroxyprogesterone hexanoate, 17α-Hydroxyprogesterone n-caproate, 3,20-Dioxopregn-4-en-17α-yl caproate, Delalutin, Depo-proluton, HPC, Hormofort, Hydroxyprogesterone caproate, Hydroxyprogesterone hexanoate, Hyproval PA, Lewntogest, NSC 17592, Neolutin, Pharlon, Primolut Depot, Procyte Depo, Proge, Progesterone caproate, Proluton Depot, Syngynon, Teralutil |
| InChI Formula | InChI=1S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1 |
| SMILES | CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] hexanoate |
| Show More |