Learn More
Hydroxyprogesterone caproate, MedChemExpress
Product Code. 16461048
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16461048
Brand: Medchem Express HYB0742
MedChemExpress Hydroxyprogesterone caproate is a synthetic, steroidal progestin; an ester derivative of 17α-hydroxyprogesterone formed from caproic acid.
Hydroxyprogesterone caproate is a synthetic, steroidal progestin
Specifications
Chemical Name or Material Hydroxyprogesterone caproate
Physical Form Solid
CAS 630-56-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C27H40O4
Synonym 17α-Hydroxyprogesterone hexanoate 17α-Hydroxyprogesterone caproate
Solubility Information DMSO : 50 mg/mL (116.66 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
SMILES CC([C@@]1(OC(CCCCC)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 428.6
Formula Weight 428.6
Percent Purity 99.68%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.