missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 3M (3N) Solution, Scharlab
Click to view available options
Quantity:
200 L
Packaging:
Metal drum
Description
- Standard volumetric solution is ready to use
- Certificate of Analysis details the factor, uncertainty level and methods used
- Traceable to the SRM of NIST
- Packaged in HDPE containers that fit into most titrators or in metal drums for bulk amounts
Specifications
Color | Colorless |
Concentration | 3mol/L |
Density | ∼1.06g/cm3 |
Packaging | Metal drum |
Traceability to NIST | Yes |
Normality | 3N |
Quantity | 200 L |
Physical Form | Liquid |
UN Number | 1789 |
Formula Weight | 36.46g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'