Learn More
Hydrochloric Acid, 0.5M, Chem-Lab
Catalog Number p-7114728
Click to view available options
Quantity:
5 L
10 L
Packaging:
PE
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
313
CHEBI:17883
hydrogen chloride
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
313
CHEBI:17883
hydrogen chloride
Cl
For laboratory use, Titrations, ASTM D-94, D-128
Concentration 18.23g HCl / 1L H2O
Density 1.01g/mL
Packaging PE
Normality 0.5N (±0.001 at 20°C)
Quantity 5 L
UN Number UN1789
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid
compliance-icons
Product Identifier
  • Hydrochloric acid 0.5 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O