Learn More
Hamilton™ HVM Small Body Valve for Modular Valve Positioner
A basic unit for the control of fluids in the laboratory.
Brand: Hamilton™ 36654
Additional Details : Weight : 0.03780kg
Description
- Physically the smallest valve
- Ideal for low pressure applications
- Loop flow path
- For use with the Modular Valve Positioner
Specifications
Loop flow path | |
Aluminum | |
14 μL | |
4-4 Model, 4 Ports |