Learn More
Hamilton™ HVM Small Body Valve for Modular Valve Positioner
A basic unit for the control of fluids in the laboratory.
Brand:  Hamilton™ 36654
Additional Details : Weight : 0.03780kg
Product Code. 11540172
  • 1700.00 NOK / Each
Estimated Shipment: 10-05-2024
Description
  • Physically the smallest valve
  • Ideal for low pressure applications
  • Loop flow path
  • For use with the Modular Valve Positioner
Specifications
Specifications
Loop flow path
Aluminum
14 μL
4-4 Model, 4 Ports