missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF323 Partial ORF (NP_112161.2, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_112161.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64288 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139596
|
Abnova™
H00064288-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129596
|
Abnova™
H00064288-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 [PubMed 12147252]).[supplied by OMIM]
Sequence: MEHLGDSRLQRDVSLDSKYRETCKRDSKAEKQQAHSTGERRHRCNECGKSFTKSSVLIEHQRIHTGEKPYECEECGKAFSRRSSLNEHRRSpecifications
NP_112161.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23407/ZNF20-Lp/ZNF310P/dJ874C20.2 | |
ZNF323 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64288 | |
ZNF323 (Human) Recombinant Protein (Q01) | |
MEHLGDSRLQRDVSLDSKYRETCKRDSKAEKQQAHSTGERRHRCNECGKSFTKSSVLIEHQRIHTGEKPYECEECGKAFSRRSSLNEHRR | |
RUO | |
ZNF323 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |