missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZMPSTE24 Partial ORF (NP_005848.2, 402 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_005848.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10269 |
Molecular Weight (g/mol) | 33.88kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142276
|
Abnova™
H00010269-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16132276
|
Abnova™
H00010269-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq]
Sequence: VLSFCLTVLSRRFEFQADAFAKKLGKAKDLYSALIKLNKDNLGFPVSDWLFSMWHYSHPPLLERLQALKTMKQHSpecifications
NP_005848.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.88kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FACE-1/FACE1/FLJ14968/STE24/Ste24p | |
ZMPSTE24 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10269 | |
ZMPSTE24 (Human) Recombinant Protein (Q01) | |
VLSFCLTVLSRRFEFQADAFAKKLGKAKDLYSALIKLNKDNLGFPVSDWLFSMWHYSHPPLLERLQALKTMKQH | |
RUO | |
ZMPSTE24 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |