Learn More
Abnova™ Human VPS11 Full-length ORF (AAH12051.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH12051.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55823 |
Molecular Weight (g/mol) | 65kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181843
|
Abnova™
H00055823-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191843
|
Abnova™
H00055823-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]
Sequence: MSEVQPDSPQGIYDTLLELRLQNWAHEKDPQVKEKLHAEAISLLKSGRFCDVFDKALVLCQMHDFQDGVLYLYEQGKLFQQIMHYHMQHEQYRQVISVCERHGEQDPSLWEQALSYFARKEEDCKEYVAAVLKHIENKNLMPPLLVVQTLAHNSTATLSVIRDYLVQKLQKQSQQIAQDELRVRRYREETTRIRQEIQELKASPKIFQKTKCSICNSALELPSVHFLCGHSFHQHCFESYSESDADCPTCLPENRKVMDMIRAQEQKRDLHDQFQHQLRCSNDSFSVIADYFGRGVFNKLTLLTDPPTARLTSSLEAGLQRDLLMHSRRGTSpecifications
AAH12051.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSEVQPDSPQGIYDTLLELRLQNWAHEKDPQVKEKLHAEAISLLKSGRFCDVFDKALVLCQMHDFQDGVLYLYEQGKLFQQIMHYHMQHEQYRQVISVCERHGEQDPSLWEQALSYFARKEEDCKEYVAAVLKHIENKNLMPPLLVVQTLAHNSTATLSVIRDYLVQKLQKQSQQIAQDELRVRRYREETTRIRQEIQELKASPKIFQKTKCSICNSALELPSVHFLCGHSFHQHCFESYSESDADCPTCLPENRKVMDMIRAQEQKRDLHDQFQHQLRCSNDSFSVIADYFGRGVFNKLTLLTDPPTARLTSSLEAGLQRDLLMHSRRGT | |
RUO | |
VPS11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55823 | |
VPS11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
END1/PEP5/RNF108/hVPS11 | |
VPS11 | |
Recombinant | |
wheat germ expression system |