missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBASH3B Partial ORF (NP_116262.2, 344 a.a. - 441 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_116262.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84959 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100987
|
Abnova™
H00084959-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190977
|
Abnova™
H00084959-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor. [provided by RefSeq]
Sequence: DGVLERRPYEDQGLGETTPLTIICQPMQPLRVNSQPGPQKRCLFVCRHGERMDVVFGKYWLSQCFDAKGRYIRTNLNMPHSLPQRSGGFRDYEKDAPISpecifications
NP_116262.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1959/MGC15437/STS-1/STS1/p70 | |
UBASH3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84959 | |
UBASH3B (Human) Recombinant Protein (Q01) | |
DGVLERRPYEDQGLGETTPLTIICQPMQPLRVNSQPGPQKRCLFVCRHGERMDVVFGKYWLSQCFDAKGRYIRTNLNMPHSLPQRSGGFRDYEKDAPI | |
RUO | |
UBASH3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |