missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRIM5 Partial ORF (NP_149023, 309 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_149023 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 85363 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181007
|
Abnova™
H00085363-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191007
|
Abnova™
H00085363-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Five alternatively spliced transcript variants for this gene have been described. However, the full length nature of two of the variants has not been determined. [provided by RefSeq]
Sequence: SCAVISEDKRQVSSPKPQIIYGARGTRYQTFVNFNYCTGILGSQSITSGKHYWEVDVSKKTAWILGVCAGFQPDAMCNIEKNENYQPKYGYWVIGLEEGVKCSAFQDSSFSpecifications
NP_149023 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RNF88/TRIM5alpha | |
TRIM5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
85363 | |
TRIM5 (Human) Recombinant Protein (Q01) | |
SCAVISEDKRQVSSPKPQIIYGARGTRYQTFVNFNYCTGILGSQSITSGKHYWEVDVSKKTAWILGVCAGFQPDAMCNIEKNENYQPKYGYWVIGLEEGVKCSAFQDSSF | |
RUO | |
TRIM5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |