Learn More
Abnova™ Human TRIM44 Full-length ORF (NP_060053.2, 1 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054765-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, namely a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. [provided by RefSeq]
Sequence: MASGVGAAFEELPHDGTCDECEPDEAPGAEEVCRECGFCYCRRHAEAHRQKFLSHHLAEYVHGSQAWTPPADGEGAGKEEAEVKVEQEREIESEAGEESESEEESESEEESETEEESEDESDEESEEDSEEEMEDEQESEAEEDNQEEGESEAEGETEAESEFDPEIEMEAERVAKRKCPDHGLDLSTYCQEDRQLICVLCPVIGAHQGHQLSTLDEAFEELRSKDSGGLKAAMIELVERLKFKSSDPKVTRDQMKMFIQQEFKKVQKVIADEEQKALHLVDIQEAMATAHVTEILADIQSHMDRLMTQMAQAKEQLDTSNESAEPKAEGDEEGPSGASEEEDTSpecifications
NP_060053.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DIPB/HSA249128/MC7/MGC3490 | |
TRIM44 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54765 | |
TRIM44 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASGVGAAFEELPHDGTCDECEPDEAPGAEEVCRECGFCYCRRHAEAHRQKFLSHHLAEYVHGSQAWTPPADGEGAGKEEAEVKVEQEREIESEAGEESESEEESESEEESETEEESEDESDEESEEDSEEEMEDEQESEAEEDNQEEGESEAEGETEAESEFDPEIEMEAERVAKRKCPDHGLDLSTYCQEDRQLICVLCPVIGAHQGHQLSTLDEAFEELRSKDSGGLKAAMIELVERLKFKSSDPKVTRDQMKMFIQQEFKKVQKVIADEEQKALHLVDIQEAMATAHVTEILADIQSHMDRLMTQMAQAKEQLDTSNESAEPKAEGDEEGPSGASEEEDT | |
RUO | |
TRIM44 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |